Learn More
Sklar Instruments Surgi-OR Hoff Towel Clamp, Sklar 6-1/4 in., Stainless Steel. EA.
SDP
Catalog No. 501184227 Shop All Sklar Instruments Products
Encompass_Preferred
Surgi-OR Hoff Towel Clamp, Sklar. 6-1/4 in., Stainless Steel. Surgi-OR Hoff Towel Clamps are a non-perforating clamp used to secure instruments and towels to the drape surrounding a surgical site. The jaws and tips are curved while the shanks and handle are flat so they can rest on the patient. The tip of this towel clamp are round with a cross-serrated surface so they grip tightly but do not penetrate the surgical drape. They also have ratcheted finger ring handles so a firm hold can be secured. This clamp should never be used on skin. Hand-crafted in Pakistan, Surgi-OR mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. Surgi-OR instruments are a cost-effective line of instruments that must meet Sklar's quality standards. This clamp is 6-1/4 inches long.
Catalog No. 50-118-4227 Supplier Sklar Instruments Supplier No. 95655
Surgi-OR Hoff Towel Clamp, Sklar. 6-1/4 in., Stainless Steel. Surgi-OR Hoff Towel Clamps are a non-perforating clamp used to secure instruments and towels to the drape surrounding a surgical site. The jaws and tips are curved while the shanks and handle are flat so they can rest on the patient. The tip of this towel clamp are round with a cross-serrated surface so they grip tightly but do not penetrate the surgical drape. They also have ratcheted finger ring handles so a firm hold can be secured. This clamp should never be used on skin. Hand-crafted in Pakistan, Surgi-OR mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. Surgi-OR instruments are a cost-effective line of instruments that must meet Sklar's quality standards. This clamp is 6-1/4 inches long.