Learn More
Description
Features:
- Urethane foam comfort
- Nitrile rubber molded to a closed cell nitrile cushion for superior comfort and performance
- Edge to edge cushion
- High heel shoe safe, welding safe, and static dissipative
- Anti-microbial treated for lifetime protection against odors and degradation
- Beveled borders
- Grease and oil proof, chemical resistant
Specifications
Specifications
| Description | Anti-microbial |
| Dimensions (L x W) | 10 x 3 ft. |
| Style | Beveled borders |
| Length (English) | 10 ft. |
| Type | Chemical-resistant Mat |
| Width (English) | 3 ft. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.