Learn More
Medchemexpress LLC HOIPIN-1 | 2470242-33-0 | 99.6% | 304.28 | 10 MG
SDP
Catalog No. 5000437465
Encompass_Preferred
HOIPIN-1 (JTP-0819958) is a selective linear ubiquitin chain assembly complex (LUBAC) inhibitor with an IC50 of 2.8 μM. It suppresses LUBAC-mediated NF-κB activation in vitro and is intended for research use only.
  • Selectively inhibits LUBAC-mediated ubiquitination
  • Suppresses NF-κB activation in vitro
  • Exhibits increased inhibitory activity with longer preincubation times
  • Inhibits expression of NF-κB target genes
Catalog No. 50-004-37465 Supplier Medchemexpress LLC Supplier No. HY12288110MG
HOIPIN-1 (JTP-0819958) is a selective linear ubiquitin chain assembly complex (LUBAC) inhibitor with an IC50 of 2.8 μM. It suppresses LUBAC-mediated NF-κB activation in vitro and is intended for research use only.
  • Selectively inhibits LUBAC-mediated ubiquitination
  • Suppresses NF-κB activation in vitro
  • Exhibits increased inhibitory activity with longer preincubation times
  • Inhibits expression of NF-κB target genes