Learn More
Grainger Sumner Hoist Alignment Tool, Steel, 1,000 lbs. Capacity, 1 ea
Catalog No. 502783816
Encompass_Preferred
The Adjust-A-Flange™ tool functions as a precision pipe fitting alignment device. Designed for professional use, this tool utilizes gravity to simplify the alignment of pipe fittings, enabling efficient one-person operation. It is constructed from durable steel and is intended to be used in conjunction with the Adjust-A-Fit™ tool for optimal functionality.
  • Features a 1,000 lbs. load capacity
  • Accommodates pipe diameters ranging from 1/8 in. (3 mm) to 24 in. (600 mm)
  • Constructed from heavy-duty steel
  • Backed by a limited lifetime warranty
Catalog No. 50-278-3816 Supplier Grainger Supplier No. 19F654
The Adjust-A-Flange™ tool functions as a precision pipe fitting alignment device. Designed for professional use, this tool utilizes gravity to simplify the alignment of pipe fittings, enabling efficient one-person operation. It is constructed from durable steel and is intended to be used in conjunction with the Adjust-A-Fit™ tool for optimal functionality.
  • Features a 1,000 lbs. load capacity
  • Accommodates pipe diameters ranging from 1/8 in. (3 mm) to 24 in. (600 mm)
  • Constructed from heavy-duty steel
  • Backed by a limited lifetime warranty