Learn More
Grainger RUD CHAIN Weld-On Hoist Ring, 3,520 lb. Capacity, Steel, 1 ea
Catalog No. 502790575
Encompass_Preferred
This weld-on hoist ring provides a robust lifting point engineered for secure material handling. Constructed from quenched and tempered steel, it offers high durability and wear resistance. The design is loadable from any direction, making it suitable for versatile lifting and lashing applications.
  • Working load limit of 3,520 lb (1,600 kg)
  • Operating temperature range from -40°F to 752°F
  • Features patented wear markings on both inside and outside surfaces
  • Designed for weld-on installation with a smaller circular fillet weld seam
  • Loadable from any direction for increased flexibility
  • Quenched and tempered for enhanced wear resistance
  • Meets DGUV test standards
Catalog No. 50-279-0575 Supplier Grainger Supplier No. 16A769
This weld-on hoist ring provides a robust lifting point engineered for secure material handling. Constructed from quenched and tempered steel, it offers high durability and wear resistance. The design is loadable from any direction, making it suitable for versatile lifting and lashing applications.
  • Working load limit of 3,520 lb (1,600 kg)
  • Operating temperature range from -40°F to 752°F
  • Features patented wear markings on both inside and outside surfaces
  • Designed for weld-on installation with a smaller circular fillet weld seam
  • Loadable from any direction for increased flexibility
  • Quenched and tempered for enhanced wear resistance
  • Meets DGUV test standards