Learn More
Grainger RUD Chain ABA Weld-On Hoist Ring, Steel, 7,050 lb WLL, 1 ea
Catalog No. 502790880
Encompass_Preferred
This weld-on lifting point is designed for secure, heavy-duty load handling in demanding industrial applications. The unit features a durable steel construction with a phosphate finish and is engineered to provide reliable performance under extreme conditions, including operating temperatures ranging from -40 °F to 752 °F.
  • Working load limit of 7,050 lb (3,200 kg)
  • Designed for weld-on mounting
  • Suitable for extreme temperature ranges
  • Durable steel construction with phosphate finish
  • Certified to DGUV test standards
Catalog No. 50-279-0880 Supplier Grainger Supplier No. 16A770
This weld-on lifting point is designed for secure, heavy-duty load handling in demanding industrial applications. The unit features a durable steel construction with a phosphate finish and is engineered to provide reliable performance under extreme conditions, including operating temperatures ranging from -40 °F to 752 °F.
  • Working load limit of 7,050 lb (3,200 kg)
  • Designed for weld-on mounting
  • Suitable for extreme temperature ranges
  • Durable steel construction with phosphate finish
  • Certified to DGUV test standards