Learn More
ATCC Staphylococcus hominis; subsp. hominis
Catalog No. 502381623
Encompass
Staphylococcus hominis subsp. hominis strain CFF 16471 is a methicillin-resistant strain that was isolated in Cleveland, Ohio, from human blood.
Catalog No. 50-238-1623 Supplier ATCC Supplier No. 51624
Staphylococcus hominis subsp. hominis strain CFF 16471 is a methicillin-resistant strain that was isolated in Cleveland, Ohio, from human blood.