Learn More
Waters Corp Holder, 20 Micron Frit, Titanium, 4 pk
Catalog No. 502392982 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This product is a holder with a 20 micron titanium frit, designed for use with ACQUITY H-Class Bio and BioQSM systems.
  • Holds frit
  • Frit pore size is 20 micron
  • Constructed from titanium
  • Supplied in a package of 4 units
  • Designed for ACQUITY H-Class Bio and BioQSM systems
  • Functions as a solvent filter
  • Suitable for UPLC systems
  • Replacement is recommended as required
Catalog No. 50-239-2982 Supplier Waters Corp Supplier No. 700005419
This product is a holder with a 20 micron titanium frit, designed for use with ACQUITY H-Class Bio and BioQSM systems.
  • Holds frit
  • Frit pore size is 20 micron
  • Constructed from titanium
  • Supplied in a package of 4 units
  • Designed for ACQUITY H-Class Bio and BioQSM systems
  • Functions as a solvent filter
  • Suitable for UPLC systems
  • Replacement is recommended as required