Learn More
Waters Corp Holder, 3/8 in. Dia, 1/2-28 Thread, 1/16 V-Check Valve
Catalog No. 502391560 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This product is a holder designed for V-check valve systems, specifically for FDM (Flow-Drift Module) check valves. It features a 3/8 in. diameter and 1/2-28 threading, making it suitable for various laboratory applications.
  • Designed for FDM check valve systems.
  • Features a 3/8 in. diameter.
  • Includes 1/2-28 threading.
  • Compatible with 1/16 in. V-check valves.
Catalog No. 50-239-1560 Supplier Waters Corp Supplier No. 700007341
This product is a holder designed for V-check valve systems, specifically for FDM (Flow-Drift Module) check valves. It features a 3/8 in. diameter and 1/2-28 threading, making it suitable for various laboratory applications.
  • Designed for FDM check valve systems.
  • Features a 3/8 in. diameter.
  • Includes 1/2-28 threading.
  • Compatible with 1/16 in. V-check valves.