Learn More
YMC America, Inc Holder for 30mmgrds -0130CCN
SDP
Catalog No. 5000876382
Encompass
Guard cartridge holder for 10 mm ID YMC guard cartridges
Catalog No. 50-008-76382 Supplier YMC America, Inc Supplier No. XPGHFSP30ID
Guard cartridge holder for 10 mm ID YMC guard cartridges