Learn More
Perkin Elmer US LLC 5-Cell Long Path length Cell Holder
Catalog No. 509042138
Encompass_Preferred
The 5-Cell Long Path length Cell Holder is compatible with the Automatic Linear Transport (B2000186) unit, an automatic cell changer for LAMBDA 20/40/20Bio/40P/25/35/45 spectrometers. The 5-Cell Long Path length Cell Holder primarily holds sample cells, but with the Reference Kit (B2000310), this holder can hold reference samples as well.
Catalog No. 50-904-2138 Supplier Perkin Elmer US LLC Supplier No. B0089439
The 5-Cell Long Path length Cell Holder is compatible with the Automatic Linear Transport (B2000186) unit, an automatic cell changer for LAMBDA 20/40/20Bio/40P/25/35/45 spectrometers. The 5-Cell Long Path length Cell Holder primarily holds sample cells, but with the Reference Kit (B2000310), this holder can hold reference samples as well.