Learn More
Med Vet International Miltex Mesh Bur & Diamond Holder, Large, 65-28
Catalog No. 502829177
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. This is a Miltex Mesh Bur & Diamond Holder. The Mesh Bur and Diamond Holder securely holds burs, diamonds or other small items preventing loss during cleaning, rinsing or sterilizing. Specifications: Large
Catalog No. 50-282-9177 Supplier Med Vet International Supplier No. MIL6528
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. This is a Miltex Mesh Bur & Diamond Holder. The Mesh Bur and Diamond Holder securely holds burs, diamonds or other small items preventing loss during cleaning, rinsing or sterilizing. Specifications: Large