Learn More
Bellco Glass, Inc. Holder for 12x24" Bag
SDP
Catalog No. 501945430
Encompass_Preferred
Place a bio-hazard bag into the sturdy wire holder and stretch the bag over the top ring. A separate hooked ring rests over the top to keep the bag in place circular shape. This keeps the bags wide open for easy access.. MADE IN THE USADiameter 185cm 7.3"Height 52cm 20.5"Material: Bright biohazard orange colored epoxy coated wire. Fits Bag: 30cm x 61cm (12 x 24") - 2200-12240
Catalog No. 50-194-5430 Supplier Bellco Glass, Inc. Supplier No. 220212024
Place a bio-hazard bag into the sturdy wire holder and stretch the bag over the top ring. A separate hooked ring rests over the top to keep the bag in place circular shape. This keeps the bags wide open for easy access.. MADE IN THE USADiameter 185cm 7.3"Height 52cm 20.5"Material: Bright biohazard orange colored epoxy coated wire. Fits Bag: 30cm x 61cm (12 x 24") - 2200-12240