Learn More
Electron Microscopy Sciences K-kits Holder
SDP
Catalog No. 501928044
Encompass_Preferred
The holder has an anodized aluminum header and a stainless steel handle. It is used with the K-kit for TEM observation of liquid samples.
Catalog No. 50-192-8044 Supplier Electron Microscopy Sciences Supplier No. K7263
The holder has an anodized aluminum header and a stainless steel handle. It is used with the K-kit for TEM observation of liquid samples.