Learn More
Perkin Elmer US LLC Brownlee NewGuard Cartridge Holder
Catalog No. NC9421090
Encompass_Preferred
This holder can be used to couple any NewGuard directly to any LC column.
Catalog No. NC9421090 Supplier Perkin Elmer US LLC Supplier No. 07150001
This holder can be used to couple any NewGuard directly to any LC column.