Learn More
Molecular Devices The QPix Microbial Colony Picker leverages best-in-class colony picking technology to alleviate bottlenecks and quickly, accurately, and efficiently screen through massive genetic libraries.
Catalog No. 5000014647
Encompass_Preferred
The QPix Microbial Colony Picker leverages best-in-class colony picking technology to alleviate bottlenecks and quickly, accurately, and efficiently screen through massive genetic libraries. The easy-to-use, intuitive software guides users through setting up colony picking runs where precision robotics pick the right colonies every time.. In addition to microbial screening, the system automates several sample preparation and plate handling processes such as transfer of bacterial liquid culture and plating on agar.
Catalog No. 50-000-14647 Supplier Molecular Devices Supplier No. X9420/220
Add to Cart
Add to Cart
Request a Quote
The QPix Microbial Colony Picker leverages best-in-class colony picking technology to alleviate bottlenecks and quickly, accurately, and efficiently screen through massive genetic libraries. The easy-to-use, intuitive software guides users through setting up colony picking runs where precision robotics pick the right colonies every time.. In addition to microbial screening, the system automates several sample preparation and plate handling processes such as transfer of bacterial liquid culture and plating on agar.