Learn More
Med Vet International McKesson Catheter Securement Device, 2.5 Inch, 172-5445-9, Each
Catalog No. 503043791
Encompass_Preferred
Catheter Stabilization Device McKesson NonSterile, 2.5 Inch Tab Made with gentle, alcohol-soluble adhesive Improves patient comfort and helps avoid complications due to Foley Provides an efficient, standardized system for securing indwelling urinary catheters May comfortably remain in place for 3-5 days without altering skin integrity Quick and easy to apply and remove Allows for repeated access to tubing Encourages proper catheter care Supports compliance with OSHA, CDC, and F315 requirements
Catalog No. 50-304-3791 Supplier Med Vet International Supplier No. 1063193EA
Catheter Stabilization Device McKesson NonSterile, 2.5 Inch Tab Made with gentle, alcohol-soluble adhesive Improves patient comfort and helps avoid complications due to Foley Provides an efficient, standardized system for securing indwelling urinary catheters May comfortably remain in place for 3-5 days without altering skin integrity Quick and easy to apply and remove Allows for repeated access to tubing Encourages proper catheter care Supports compliance with OSHA, CDC, and F315 requirements