Learn More
Cole-Parmer H-B Instrument B61400-7300 Stainless Steel Clamping Thermometer Holder for 3.5 to 14.5mm Thermometers
Catalog No. 50200177 Shop All Cole-Parmer Products
Encompass_Preferred
Hold your thermometer firmly to the side of any beaker with this handy clip. Foam rubber holder has five holes ranging from 4 to 14 mm in diameter to hold almost any size thermometer. Constructed of steel and rubber for long life.
Catalog No. 50-200-177 Supplier Cole-Parmer Supplier No. 0812090
Hold your thermometer firmly to the side of any beaker with this handy clip. Foam rubber holder has five holes ranging from 4 to 14 mm in diameter to hold almost any size thermometer. Constructed of steel and rubber for long life.