Learn More
Med Vet International Mayo-Hegar Needle Holder, 8-in., Fenestrated Jaws, Von Klaus German Surgical Steel
Catalog No. 502092274
Encompass_Preferred
Mayo-Hegar needle holders are used widely in general surgery and are meant to hold medium to large heavy suture needles. The Von Klaus line of instruments is made of high quality German surgical steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-209-2274 Supplier Med Vet International Supplier No. VK1101820
Mayo-Hegar needle holders are used widely in general surgery and are meant to hold medium to large heavy suture needles. The Von Klaus line of instruments is made of high quality German surgical steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.