Learn More
Med Vet International 5.5-in Olsen-Hegar Tungsten Carbide Needle Holder with Integrated Scissors, Serrated, Von Klas
Supplier:  Med Vet International VK132160
Encompass_Preferred
This instrument is serrated with integrated scissors and are generally used with sutures. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-209-2279