Learn More
Med Vet International Olsen-Hegar Needle Holder/Scissors, 6.5-in, Serrated Jaws, Von Klaus German Surgical Steel
Catalog No. 502092284
Encompass_Preferred
This Von Klaüs German-grade Olsen-Hegar needle holder is a ratcheted, finger ring instrument that also has built-in scissors. It is used with intermediate weight sutures, commonly in plastic surgery procedures. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-209-2284 Supplier Med Vet International Supplier No. VK1106017
This Von Klaüs German-grade Olsen-Hegar needle holder is a ratcheted, finger ring instrument that also has built-in scissors. It is used with intermediate weight sutures, commonly in plastic surgery procedures. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.