Learn More
Electron Microscopy Sciences MAC Holders for 12 Standards for JXA 733
SDP
Catalog No. 5028772
Encompass_Preferred
Special holders for use with MAC reference standards. JXA 733 Standard Holder in brass; holds up to 12 standards of your choice.
Catalog No. 50-287-72 Supplier Electron Microscopy Sciences Supplier No. 8007303
Special holders for use with MAC reference standards. JXA 733 Standard Holder in brass; holds up to 12 standards of your choice.