Learn More
Electron Microscopy Sciences MAC Holders for 21 Standards for JSM 35
SDP
Supplier:  Electron Microscopy Sciences 8007301
Encompass_Preferred
Special holders for use with MAC reference standards. JSM 35 Standard Holder in brass, holds up to 21 standards of your choice.
Catalog No. 50-287-74