Learn More
Electron Microscopy Sciences MAC Holders for 3-5 Standards for JEOL Temscan
SDP
Catalog No. 5028773
Encompass_Preferred
Special holders for use with MAC reference standards. JEOL Temscan bulk Standard Holder made from plastic; holds 3-5 standards, fitted in a JEOL BR1 holder.
Catalog No. 50-287-73 Supplier Electron Microscopy Sciences Supplier No. 8007304
Special holders for use with MAC reference standards. JEOL Temscan bulk Standard Holder made from plastic; holds 3-5 standards, fitted in a JEOL BR1 holder.