Learn More
Electron Microscopy Sciences MAC Holders for 7 Standards for JEOL 100cx
SDP
Catalog No. 5028771
Encompass_Preferred
Special holders for use with MAC reference standards. JEOL 100CX Standard Holder in brass for use in the STEM mode, holds up to 7 standards of your choice.
Catalog No. 50-287-71 Supplier Electron Microscopy Sciences Supplier No. 8007302
Special holders for use with MAC reference standards. JEOL 100CX Standard Holder in brass for use in the STEM mode, holds up to 7 standards of your choice.