Learn More
VITROLIFE INC Holding Pipettes
SDP
Catalog No. NC3356088
Encompass
Pipette with a rounded tip and polished opening.
To hold the oocyte or embryo when performing ICSI or other micromanipulation procedures.
For use with a micromanipulator.
Store in +2 to +30°C
This item is not returnable. View return policy
Catalog No. NC3356088 Supplier VITROLIFE INC Supplier No. 15331
Pipette with a rounded tip and polished opening.
To hold the oocyte or embryo when performing ICSI or other micromanipulation procedures.
For use with a micromanipulator.
Store in +2 to +30°C