Learn More
MSC SPI Mechanical Hole Micrometer, 0.5000 to 0.6500 in. Range, Solid Carbide, 1 ea
Catalog No. 5000513407
Encompass
This mechanical three-point internal hole micrometer is designed for the precise direct measurement of bore diameters within a range of 0.5000 in. to 0.6500 in. It features a robust, self-aligning and self-centering design that ensures accurate, repeatable readings in industrial metalworking applications.
  • Solid carbide measuring anvils for durability
  • Ratchet stop provides uniform measuring pressure for consistent accuracy
  • Suitable for measuring within 0.050 in. of the bottom of blind holes
  • Feelers taper toward the back to allow positive reading of the front edge
  • Measuring depth of 2.36 in.
Catalog No. 50-005-13407 Supplier MSC Supplier No. 00342915
This mechanical three-point internal hole micrometer is designed for the precise direct measurement of bore diameters within a range of 0.5000 in. to 0.6500 in. It features a robust, self-aligning and self-centering design that ensures accurate, repeatable readings in industrial metalworking applications.
  • Solid carbide measuring anvils for durability
  • Ratchet stop provides uniform measuring pressure for consistent accuracy
  • Suitable for measuring within 0.050 in. of the bottom of blind holes
  • Feelers taper toward the back to allow positive reading of the front edge
  • Measuring depth of 2.36 in.