Learn More
Mitegen Au-Flat TEM Grid, 2µm Hole Pattern on 200 Gold Mesh, For Cryo-EM Sample Preparation,
SDP
Catalog No. NC3361404
Encompass
The Au-Flat TEM Grid features a precise 2 µm hole pattern on a robust 200 gold mesh, making it an essential component for high-resolution cryo-electron microscopy (cryo-EM) sample preparation. Crafted from high-quality gold, this grid offers excellent conductivity and stability, which are critical for optimal imaging performance and minimizing sample drift in cryogenic environments. The uniform hole pattern facilitates consistent ice thickness and particle distribution, crucial for achieving high-quality data. Designed for reliability and ease of handling, these grids support a streamlined workflow for vitrification and subsequent imaging. They provide a stable and optically clear substrate for delicate biological specimens, contributing to enhanced resolution and reproducibility in structural biology research. This product is supplied in a convenient box of 50 grids, ensuring an ample supply for demanding, high-volume experimental needs in cryo-EM.
Catalog No. NC3361404 Supplier Mitegen Supplier No. MCEMAUFT22250
The Au-Flat TEM Grid features a precise 2 µm hole pattern on a robust 200 gold mesh, making it an essential component for high-resolution cryo-electron microscopy (cryo-EM) sample preparation. Crafted from high-quality gold, this grid offers excellent conductivity and stability, which are critical for optimal imaging performance and minimizing sample drift in cryogenic environments. The uniform hole pattern facilitates consistent ice thickness and particle distribution, crucial for achieving high-quality data. Designed for reliability and ease of handling, these grids support a streamlined workflow for vitrification and subsequent imaging. They provide a stable and optically clear substrate for delicate biological specimens, contributing to enhanced resolution and reproducibility in structural biology research. This product is supplied in a convenient box of 50 grids, ensuring an ample supply for demanding, high-volume experimental needs in cryo-EM.