Learn More
MSC M.K. MORSE Hole Saw, Bi-Metal, 2 in, 1 ea
Supplier:  MSC 32552689
Encompass
This bi-metal, toothed edge hole saw is designed for various demanding applications. It features a 2-inch saw diameter and a 1-15/16 inch cutting depth, making it suitable for drilling precise holes in a wide range of materials.
  • Bi-metal construction for durability and extended life
  • Toothed edge for efficient and clean cutting
  • Two inch saw diameter for precise hole cutting
  • One and 15/16 inch cutting depth suitable for various material thicknesses
  • Variable pitch pattern for smooth operation
  • M42 material grade for enhanced performance
  • Compatible with a wide range of materials including cement board, steel, brick, and cast iron
  • Made in the United States
Catalog No. 50-192-0199