Learn More
Electron Microscopy Sciences EMS Holey Carbon Film 50 µm holes on 200 mesh Copper
SDP
Catalog No. 5024445
Encompass_Preferred
A thin film of carbon with round holes. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. NOTE: All of our film is laid on the shiny side of the grid.
Grid: Copper
Holes: 50 µm
Catalog No. 50-244-45 Supplier Electron Microscopy Sciences Supplier No. HC200CU
A thin film of carbon with round holes. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. NOTE: All of our film is laid on the shiny side of the grid.
Grid: Copper
Holes: 50 µm