Learn More
Electron Microscopy Sciences EMS Holey Carbon Film 4-6 nm thick on 300 mesh Nickel Grid
SDP
Catalog No. 501931368
Encompass_Preferred
A thin film of carbon with round holes. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. NOTE: All of our film is laid on the shiny side of the grid.
Grid: Nickel
Thickness: 4-6 nm
Catalog No. 50-193-1368 Supplier Electron Microscopy Sciences Supplier No. HC300NIUL
A thin film of carbon with round holes. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. NOTE: All of our film is laid on the shiny side of the grid.
Grid: Nickel
Thickness: 4-6 nm