Learn More
Electron Microscopy Sciences EMS Holey Carbon Film 150 µm holes on 200 mesh Gold
SDP
Catalog No. 501902784
Encompass_Preferred
A thin film of carbon with round holes. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. NOTE: All of our film is laid on the shiny side of the grid.
Grid: Gold
Holes: 150 µm
Catalog No. 50-190-2784 Supplier Electron Microscopy Sciences Supplier No. HC200AU150
A thin film of carbon with round holes. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. NOTE: All of our film is laid on the shiny side of the grid.
Grid: Gold
Holes: 150 µm