Learn More
Electron Microscopy Sciences Quantifoil Holey SiO2 Film R 1.2/1.3, Nickel 300 mesh Grid
SDP
Catalog No. 501927691
Encompass_Preferred
QUANTIFOIL is a perforated Silicon dioxide SiO2 support foil with pre-defined hole size, shape and arrangement. Used in cryo-electron microscopy at magnifications of 50,000x and higher. ~1.2 µm holes with 2.5 µm period.
Catalog No. 50-192-7691 Supplier Electron Microscopy Sciences Supplier No. Q3100NR1.3S
QUANTIFOIL is a perforated Silicon dioxide SiO2 support foil with pre-defined hole size, shape and arrangement. Used in cryo-electron microscopy at magnifications of 50,000x and higher. ~1.2 µm holes with 2.5 µm period.