Learn More
Electron Microscopy Sciences Quantifoil Holey SiO2 Film R 1.2/21.2, Nickel 200 mesh Grid
SDP
Catalog No. 501927696
Encompass_Preferred
QUANTIFOIL is a perforated Silicon dioxide SiO2 support foil with pre-defined hole size, shape and arrangement. Used in cryo-electron microscopy at magnifications of 50,000x and higher.. 1.2 µm hole and 21.2 µm period.
Catalog No. 50-192-7696 Supplier Electron Microscopy Sciences Supplier No. Q2100NR21.2S
QUANTIFOIL is a perforated Silicon dioxide SiO2 support foil with pre-defined hole size, shape and arrangement. Used in cryo-electron microscopy at magnifications of 50,000x and higher.. 1.2 µm hole and 21.2 µm period.