Learn More
Electron Microscopy Sciences Quantifoil Holey SiO2 Film R 1/4, Copper 400 mesh Grid
SDP
Catalog No. 501927680
Encompass_Preferred
QUANTIFOIL is a perforated Silicon dioxide SiO2 support foil with pre-defined hole size, shape and arrangement. Used in cryo-electron microscopy where the carbon film should be outside the frame of the image. 1 µm hole and 5 µm period.
Catalog No. 50-192-7680 Supplier Electron Microscopy Sciences Supplier No. Q450CR14S
QUANTIFOIL is a perforated Silicon dioxide SiO2 support foil with pre-defined hole size, shape and arrangement. Used in cryo-electron microscopy where the carbon film should be outside the frame of the image. 1 µm hole and 5 µm period.