Learn More
Electron Microscopy Sciences Quantifoil Holey SiO2 Film R 2/2, Copper 300 mesh Grid
SDP
Catalog No. 501927700
Encompass_Preferred
QUANTIFOIL is a perforated Silicon dioxide SiO2 support foil with pre-defined hole size, shape and arrangement. Used in cryo-electron microscopy at magnifications between 30,000x and 40,000x. 2 µm hole and 4 µm period.
Catalog No. 50-192-7700 Supplier Electron Microscopy Sciences Supplier No. Q3100CR2S
QUANTIFOIL is a perforated Silicon dioxide SiO2 support foil with pre-defined hole size, shape and arrangement. Used in cryo-electron microscopy at magnifications between 30,000x and 40,000x. 2 µm hole and 4 µm period.