Learn More
Electron Microscopy Sciences Quantifoil Holey SiO2 Film R 2/4, Copper 200 mesh Grid
SDP
Catalog No. 501927708
Encompass_Preferred
QUANTIFOIL is a perforated Silicon dioxide SiO2 support foil with pre-defined hole size, shape and arrangement. Used in cryo-electron microscopy where the carbon film should be outside the frame of the image. 2 µm hole and 6 µm period.
Catalog No. 50-192-7708 Supplier Electron Microscopy Sciences Supplier No. Q2100CR4S
QUANTIFOIL is a perforated Silicon dioxide SiO2 support foil with pre-defined hole size, shape and arrangement. Used in cryo-electron microscopy where the carbon film should be outside the frame of the image. 2 µm hole and 6 µm period.