Learn More
Ace Glass, Inc. Stopcock plug, 19/38, hollow, inlet feed, Buchi 40628 &00677
SDP
Catalog No. 50977952
Encompass
19/38 hollow stopcock plug with inlet feed for Buchi rotary evaporators 40628 and 00677.
Catalog No. 50-977-952 Supplier Ace Glass, Inc. Supplier No. 1329510
19/38 hollow stopcock plug with inlet feed for Buchi rotary evaporators 40628 and 00677.