Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ Spherical Penny Head Stopper
GREENER_CHOICESDP

Catalog No. 31501893 Shop All Kemtech Products
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
31-501-893 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 31-501-893 Supplier Synthware™ Supplier No. S450035
Add to Cart
Add to Cart

35/20 Ball Joint Penny Head Stopper

Specifications

For Use With (Application) Organic Synthesis
Description Heavy-wall
Quantity 1 Ea.
Color Clear
Material Borosilicate Glass
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.