Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ Hollow Penny Head Stopper
GREENER_CHOICESDP

Catalog No. 31501875 Shop All Kemtech Products
Catalog No. 31-501-875 Supplier Synthware™ Supplier No. S404550
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Heavy wall hollow penny head stopper

Specifications

Autoclavable Autoclavable
Diameter (Metric) Outer 45 mm
Product Type Hollow Pennyhead Stopper
Shape Cylindrical
Quantity 1 Ea.
Color Clear
Material Borosilicate Glass
Description Heavy-wall
For Use With (Application) Organic Synthesis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.