Specifications
| For Use With (Application) | Organic Synthesis |
| Material | Borosilicate Glass |
| Product Type | Stopper with #15 O-Ring Joint |
| Quantity | 1 Ea. |
| Standard Taper Joint No. | 14/20 Bottom, 24/40 Top |
| Thread Size | #15 O-ring |