Learn More
Strem, An Ascensus Company CAS# 14914-84-2. 10g. Holmium(III) chloride hexahydrate (99.9%-Ho) (REO). MFCD00149739
SDP
Catalog No. 5090116146
Encompass
CAS# 14914-84-2. 10g. Holmium(III) chloride hexahydrate (99.9%-Ho) (REO). MFCD00149739. Molecular Weight: 271.29 (379.38). Molecular Formula: HoCl3∙6H2O. Color/form: pink xtl. Strem# 93-6722. http://www.strem.com/catalog/v/93-6722/
Catalog No. 50-901-16146 Supplier Strem, An Ascensus Company Supplier No. 93672210G
CAS# 14914-84-2. 10g. Holmium(III) chloride hexahydrate (99.9%-Ho) (REO). MFCD00149739. Molecular Weight: 271.29 (379.38). Molecular Formula: HoCl3∙6H2O. Color/form: pink xtl. Strem# 93-6722. http://www.strem.com/catalog/v/93-6722/