Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Holmium(III) fluoride, anhydrous, 99.9% (REO)

Catalog No. AA4141309
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4141309 10 g
AA4141304 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4141309 Supplier Thermo Scientific Chemicals Supplier No. 04141309
Add to Cart

CAS: 13760-78-6 | F3Ho | 221.93 g/mol

For vacuum deposition

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13760-78-6
Molecular Formula F3Ho
Molecular Weight (g/mol) 221.93
MDL Number MFCD00016142
InChI Key FDIFPFNHNADKFC-UHFFFAOYSA-K
IUPAC Name holmium(3+) trifluoride
SMILES [F-].[F-].[F-].[Ho+3]

Specifications

Melting Point 1143°C
Physical Form Pieces
Quantity 10 g
UN Number UN3288
Formula Weight 221.93
Percent Purity 99.9%
Packaging Glass bottle
Sensitivity Hygroscopic
Specific Gravity 7.6
Density 7.6
Chemical Name or Material Holmium(III) fluoride
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 237-352-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.