Alfa Aesar™ Holmium(III) nitrate pentahydrate, REacton™, 99.9% (REO)

Manufacturer: Alfa Aesar™ 1458818

Quantity 50g

 View more versions of this product

Catalog No. AA1458818

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14483-18-2
    Formula Weight 441.02 (350.94 Anhydrous)
    MDL Number MFCD00149737
    UN Number UN1477
    Chemical Name or Material Holmium(III) nitrate pentahydrate
    Grade Reacton™

    Specifications

    Formula Weight 441.02 (350.94 Anhydrous)
    Health Hazard 3 GHS P Statement
    P221-P210-P220-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Keep/Store away from clothing/combustible materials.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Crystalline Aggregates
    Quantity 50g
    Sensitivity Hygroscopic
    MDL Number MFCD00149737
    CAS 14483-18-2
    TSCA TSCA
    Odor Odorless
    UN Number UN1477
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H272-H315-H319-H335
    May intensify fire
    oxidizer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.
    Chemical Name or Material Holmium(III) nitrate pentahydrate
    Assay 99.9%, (REO)
    Grade Reacton™

    GHS H Statement
    H272-H315-H319-H335
    May intensify fire
    oxidizer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P221-P210-P220-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Keep/Store away from clothing/combustible materials.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit