Learn More
Strem, An Ascensus Company CAS# 12055-62-8. 5g. Holmium(III) oxide (99.9%-Ho) (REO). MFCD00011053
SDP
Catalog No. 5090116141
Encompass
CAS# 12055-62-8. 5g. Holmium(III) oxide (99.9%-Ho) (REO). MFCD00011053. Molecular Weight: 377.86. Molecular Formula: Ho2O3. Color/form: off-white to pink pwdr. Strem# 93-6713. http://www.strem.com/catalog/v/93-6713/
Catalog No. 50-901-16141 Supplier Strem, An Ascensus Company Supplier No. 9367135G
CAS# 12055-62-8. 5g. Holmium(III) oxide (99.9%-Ho) (REO). MFCD00011053. Molecular Weight: 377.86. Molecular Formula: Ho2O3. Color/form: off-white to pink pwdr. Strem# 93-6713. http://www.strem.com/catalog/v/93-6713/