Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Holmium(III) oxide, REacton™, 99.99% (REO)

Catalog No. AA1128118
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1128118 50 g
AA1128109 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1128118 Supplier Thermo Scientific Chemicals Supplier No. 01128118
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 12055-62-8 | Ho2O3 | 377.86 g/mol

Holmium(III) oxide iacts as colorants which is used for cubic zirconia and glass, as a calibration standard for optical spectrophotometers, as a specialty catalyst, phosphor and a laser material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12055-62-8
Molecular Formula Ho2O3
Molecular Weight (g/mol) 377.86
MDL Number MFCD00011053
InChI Key OWCYYNSBGXMRQN-UHFFFAOYSA-N
IUPAC Name diholmium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Ho+3].[Ho+3]

Specifications

Melting Point 2360°C
Color Beige
Physical Form Powder
Quantity 50 g
Odor Odorless
Assay Percent Range 99.99% (REO)
Solubility Information Insoluble in water. Soluble in perchloric acids.
Formula Weight 377.86
Chemical Name or Material Holmium(III) oxide
EINECSNumber 235-015-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.