Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Holmium(III) trifluoromethanesulfonate hydrate, 98%, Thermo Scientific Chemicals

Catalog No. p-7051028
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4030709 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4030709 Supplier Thermo Scientific Chemicals Supplier No. 04030709
Only null left
Add to Cart

It is a water-tolerant Lewis acid used in the Aldol reaction of silyl enol ethers with aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139177-63-2
Molecular Formula C3F9HoO9S3
Molecular Weight (g/mol) 612.12
MDL Number MFCD00209590
InChI Key DBPDCYACEYLEMY-UHFFFAOYSA-K
Synonym holmium iii trifluoromethanesulfonate, holmium triflate, holmium iii trifluoromethanesulphonate, trifluoromethanesulfonic acid holmium iii salt, methanesulfonic acid,1,1,1-trifluoro-, holmium 3+ salt 3:1, holmium trifluoromethanesulfonate, holmium iii triflate, ho otf 3, acmc-1bwo7
PubChem CID 22482917
IUPAC Name holmium(3+);trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Ho+3]

Specifications

Physical Form Powder
Quantity 10 g
Formula Weight 612.13 (Anhydrous)
Percent Purity 98%
Chemical Name or Material Holmium(III) trifluoromethanesulfonate hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.