Learn More
Thermo Scientific™ Holmium Oxide Solution Verification Standard
Catalog No. 14386449 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Holmium Oxide in Perchloric Acid Solution
Catalog No. Type
14-386-449 Holmium Oxide in Perchloric Acid Solution
1 options
This standard is qualified for use on instruments having a spectral bandwidth of <3nm.
Catalog No. 14-386-449 Supplier Thermo Scientific™ Supplier No. 222226400
Add to Cart
Add to Cart
This standard is qualified for use on instruments having a spectral bandwidth of <3nm.
Thermo Scientific™ NIST™-traceable Holmium Oxide Solution Standard verifies wavelength accuracy of UV-Visible spectrophotometers.
Thermo Scientific NIST™-traceable Holmium Oxide Solution Standard verifies wavelength accuracy of UV-Visible spectrophotometers.
Holmium Oxide in Perchloric Acid Solution is a preferred standard for verifying the wavelength accuracy of a spectrophotometer according to international pharmacopoeia guidelines.
  • Permanently sealed in a quartz cuvette allowing it to be used into the deep UV range
  • Exhibits sharp, stable peaks within the range of 219 to 650nm—can be easily correlated to wavelengths of the peak maxima
  • Readings observed for each peak are verified against expected values on the certificate supplied with the standard
  • Readings not within the expected range may be indicative of an instrument performance error
This standard is qualified for use on instruments having a spectral bandwidth of <3nm.
TRUSTED_SUSTAINABILITY
Specifications
Calibration NIST/NPL Traceable
Description Wavelength Standard; Verification Standard
Type Holmium Oxide in Perchloric Acid Solution