Learn More
Athens Research And Technology Inc HOLO TRANSFERRIN HU 100MG
Supplier:  Athens Research And Technology Inc 1616032001HOLO100MG
Encompass_Preferred
Transferrin (HOLO), Human Plasma: 100 MG; Lyophilized from 20 mM Na phosphate, pH 7.4, with 150 mM NaCl.; MW 80,000; Iron Content: > = 1100UG/GR; Purity: > =95% by SDS-PAGE. Native; Purified from human plasma in the Athens Research & Tech lab.
Catalog No. 50-489-756