Learn More
TARGETMOL CHEMICALS INC HOLOMYCIN 5MG
SDP
Catalog No. 502579274
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Holomycin (N-Demethylthiolutin) is a secondary metabolite of dithiopyrrolidone. HolomycinHolomycin an antibiotic secondary metabolite is required for the biofilm formation of natural photogenic Bacillus galatheae S2753 and is also a broad-spectrum antibiotic. holomycin promotes biofilm formation of S2753 through its alene-disthionyl group. Holomycin has antitumor activity and is involved in RNA synthesis. purity: 98%
Catalog No. 50-257-9274 Supplier TARGETMOL CHEMICALS INC Supplier No. T677315MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Holomycin (N-Demethylthiolutin) is a secondary metabolite of dithiopyrrolidone. HolomycinHolomycin an antibiotic secondary metabolite is required for the biofilm formation of natural photogenic Bacillus galatheae S2753 and is also a broad-spectrum antibiotic. holomycin promotes biofilm formation of S2753 through its alene-disthionyl group. Holomycin has antitumor activity and is involved in RNA synthesis. purity: 98%